4-(Dimethylamino)-N-[7-(hydroxyamino)-7-oxoheptyl]benzamide ≥97.0% (by HPLC)
: TCI
:251456-60-7
:
:C₁₆H₂₅N₃O₃
:MFCD03453554
:307.39 g/mol
:Refrigerator
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 97.0 area%
- Elemental analysis (Nitrogen):13.2 to 13.8 %
- Melting point:150.0 to 154.0 deg-C
Frequently Bought Together