2,9-Dichloro-1,10-phenanthroline ≥97.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:29176-55-4
:
Formula:C₁₂H₆Cl₂N₂
MDL Number:MFCD02084186
:249.09 g/mol
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 97.0 %
- Purity (Nonaqueous Titration):min. 97.0 %
- Melting point:233.0 to 237.0 deg-C
Specification Test Results
Frequently Bought Together