9(10)-Dehydronandrolone ≥97.0% (by HPLC)
: TCI
:6218-29-7
:
:C₁₈H₂₄O₂
:MFCD08457892
:272.39 g/mol
:190°C
- Appearance:Light yellow to Yellow to Orange powder to crystal
- Purity (HPLC):min. 97.0 area%
- Specific rotation [a]20/D:-310 to -330 deg (C=0.5, MeOH)
Frequently Bought Together