Dibenzothiophene-5,5-dioxide ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:1016-05-3
:
Formula:C₁₂H₈O₂S
MDL Number:MFCD00004970
:216.25 g/mol
EINECS:213-805-9
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:234.0 to 238.0 deg-C
Specification Test Results
Frequently Bought Together