(S)-O-Desmethylraclopride ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:119670-11-0
:
Formula:C₁₄H₁₈Cl₂N₂O₃
MDL Number:MFCD02179385
:333.21 g/mol
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:221.0 to 225.0 deg-C
- Specific rotation [a]20/D:-30.0 to -33.0 deg (C=1,DMSO)
Specification Test Results
Frequently Bought Together