Direct red 80 for biochemical research
Catalog # TCIAD4132-5G
Supplier: TCI
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:Red to Dark blue to Black powder to crystal
- Lambda max.:524.0 to 528.0 nm (H2O)
- Absorbance (E1%1cm):min. 350 (H2O, 524.0 to 528.0 nm)
- Collagen detection test:to pass test
Specification Test Results
About this item
:
:1373.051 g/mol
Merck Index:14,08548
EINECS:220-027-3
CAS Number:2610-10-8
Formula:C₄₅H₂₆N₁₀Na₆O₂₁S₆
MDL Number:MFCD00054389