
Dimethyldioctadecylammonium Iodide ≥98.0% (by total nitrogen basis)
Catalog # TCIAD4124-5G
Supplier: TCI
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to Light yellow to Light red powder to crystal
- Purity (with Total Nitrogen):min. 98.0 %
- Melting point:152.0 to 156.0 deg-C
Specification Test Results
About this item
:
:677.97 g/mol
:GHS07
CAS Number:7206-39-5
Formula:C₃₈H₈₀IN