

2,3-Diaminonaphthalene ≥98.0% (by GC, titration analysis) for biochemical research
Catalogus # TCIAD4101-100MG
Supplier: TCI
CAS Number:
Specifications
- Pk:100 mg
- Seq:2440
Specifications
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
- Nitrite ion detection test(fluorescence method):to pass test
Specification Test Results
About this item
:
:158.2 g/mol
EINECS:212-241-0
:GHS07
:GHS08
Storage Temperature:Refrigerator
CAS Number:771-97-1
Formula:C₁₀H₁₀N₂
MDL Number:MFCD00004116