
(3,4-Dimethoxybenzyl)triphenylphosphonium bromide ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAD4072-5G
Supplier: TCI
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:239.0 to 243.0 deg-C
Specification Test Results
About this item
:
:493.38 g/mol
:GHS07
CAS Number:70219-09-9
Formula:C₂₇H₂₆BrO₂P
MDL Number:MFCD20257911