Order Entry
Netherlands
ContactUsLinkComponent
6,6'-Dibromo-2,2'-bipyridyl ≥95.0% (by GC)
Irriterend
6,6'-Dibromo-2,2'-bipyridyl ≥95.0% (by GC)
Leverancier:  TCI
Certificaten
Irriterend
6,6'-Dibromo-2,2'-bipyridyl ≥95.0% (by GC)
Leverancier:  TCI

Specificatie testresultaten

  • Appearance:
    White to Orange to Green powder to crystal
  • Purity (GC):
    min. 95.0 %
  • Melting point:
    223.0 to 227.0 deg-C

Specificatie testresultaten

Gerelateerde informatie

Frequently Bought Together