Order Entry
Netherlands
ContactUsLinkComponent
(-)-(5aS,10bR)-5a,10b-Dihydro-2-(2,4,6-trimethylphenyl)-4H,6H-indeno[2,1-b][1,2,4]triazolo[4,3-d][1,4]oxazinium chloride monohydrate ≥98.0% (by HPLC)
(-)-(5aS,10bR)-5a,10b-Dihydro-2-(2,4,6-trimethylphenyl)-4H,6H-indeno[2,1-b][1,2,4]triazolo[4,3-d][1,4]oxazinium chloride monohydrate ≥98.0% (by HPLC)
Catalog # TCIAD3984-100MG
Supplier:  TCI
CAS Number:  
undefined
(-)-(5aS,10bR)-5a,10b-Dihydro-2-(2,4,6-trimethylphenyl)-4H,6H-indeno[2,1-b][1,2,4]triazolo[4,3-d][1,4]oxazinium chloride monohydrate ≥98.0% (by HPLC)
Catalog # TCIAD3984-100MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Seq:
    2440

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Elemental analysis (Nitrogen):
    10.50 to 11.50 %
  • Specific rotation [a]20/D:
    -156 to -160 deg (C=1, CHCl3)

Specification Test Results

About this item

 :
 :367.88 g/mol
CAS Number:919102-70-8
Formula:C₂₁H₂₂ClN₃O
MDL Number:MFCD10566990