(+)-(5aR,10bS)-5a,10b-Dihydro-2-(2,4,6-trimethylphenyl)-4H,6H-indeno[2,1-b][1,2,4]triazolo[4,3-d][1,4]oxazinium chloride monohydrate ≥98.0% (by HPLC)
Catalog # TCIAD3983-100MG
Supplier: TCI
CAS Number:
Specifications
- Pk:100 mg
- Seq:2440
Specifications
Specification Test Results
- Appearance:White to Gray to Red powder to crystal
- Purity (HPLC):min. 98.0 area%
- Elemental analysis (Nitrogen):10.40 to 11.20 %
- Specific rotation [a]20/D:+155 to +163 deg (C=1, CHCl3)
Specification Test Results
About this item
:
:367.87 g/mol
CAS Number:903571-02-8
:MFCD09787522
Formula:C₂₁H₂₂ClN₃O
MDL Number:MFCD10566989