N,N'-Di-1-naphthyl-N,N'-diphenylbenzidine ≥98.0% (by HPLC, total nitrogen), purified by sublimation
Supplier: TCI
Certificates
About this item
CAS Number:123847-85-8
:
Formula:C₄₄H₃₂N₂
MDL Number:MFCD03093246
:588.75 g/mol
Specification Test Results
- Appearance:White to Yellow to Green powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
Specification Test Results
Frequently Bought Together