2,6-Dibromobenzo[1,2-b:4,5-b']dithiophene ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:909280-97-3
:
Formula:C₁₀H₄Br₂S₂
:348.07 g/mol
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
Specification Test Results
Frequently Bought Together