
2,7-Dibromo-9,9-dihexylfluorene ≥98.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:189367-54-2
:
Formula:C₂₅H₃₂Br₂
MDL Number:MFCD03427215
:492.34 g/mol
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Melting point:66.0 to 70.0 deg-C
Specification Test Results
Frequently Bought Together