
2,7-Dibromocarbazole ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI
Certificates
About this item
CAS Number:136630-39-2
:
Formula:C₁₂H₇Br₂N
MDL Number:MFCD09033507
:325.00 g/mol
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
- Melting point:230.0 to 234.0 deg-C
Specification Test Results
Frequently Bought Together