1,2-Distearoyl-sn-glycero-3-phosphocholine ≥95.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:816-94-4
:
Formula:C₄₄H₈₈NO₈P
MDL Number:MFCD00036905
:790.16 g/mol
Melting Pt:236°C
Storage Temperature:Freezer
EINECS:212-440-2
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC) (CAD):min. 95.0 area%
- Purity (Nonaqueous Titration):min. 95.0 %
- Specific rotation [a]20/D:+5.0 to +7.0 deg (C=2、CHCl3)
Specification Test Results
Frequently Bought Together