
Diphenyl-4-bromo-ɑ-chlorobenzylphosphonate ≥95.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:189099-56-7
:
Formula:C₁₉H₁₅BrClO₃P
:437.65 g/mol
Melting Pt:107°C
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 90.0 area%
- Purity (Argentmetric Titration):min. 95.0 %
Specification Test Results
Frequently Bought Together