
4,7-Dihydroxy-1,10-phenanthroline ≥98.0% (by HPLC)
Catalog # TCIAD3869-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Gray to Brown powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 97.0 %
Specification Test Results
About this item
:
:212.21 g/mol
EINECS:223-493-6
:GHS07
Storage Temperature:Freezer
CAS Number:3922-40-5
Formula:C₁₂H₈N₂O₂
MDL Number:MFCD00004975