Order Entry
Netherlands
ContactUsLinkComponent
 
 
Diethylene glycol-2-bromoethyl methyl ether ≥94.0% (by GC)
 :  TCI
 
 
Diethylene glycol-2-bromoethyl methyl ether ≥94.0% (by GC)
 :  TCI

 

  • Appearance:
    Colorless to Brown clear liquid
  • Purity (GC):
    min. 94.0 %

 

Frequently Bought Together