Order Entry
Netherlands
ContactUsLinkComponent
9,9-Dimethylfluorene ≥99.0% (by GC), purified by sublimation
9,9-Dimethylfluorene ≥99.0% (by GC), purified by sublimation
  TCIAD3810-1G
 :  TCI
 :  
undefined
9,9-Dimethylfluorene ≥99.0% (by GC), purified by sublimation
  TCIAD3810-1G
 :  TCI
 :  

 

  • Pk:
    1 g
  • Seq:
    3047

 

 

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 99.0 %
  • Melting point:
    95.0 to 99.0 deg-C

 

 

 :
 :194.28 g/mol
 :287°C
 :4569-45-3
 :C₁₅H₁₄
 :MFCD00114670