Order Entry
Netherlands
ContactUsLinkComponent
1,1-Dimethyl-3-[3-(trifluoromethyl)phenyl]urea ≥97.0% (by total nitrogen basis)
Irritant
1,1-Dimethyl-3-[3-(trifluoromethyl)phenyl]urea ≥97.0% (by total nitrogen basis)
Supplier:  TCI
Certificates
Irritant
1,1-Dimethyl-3-[3-(trifluoromethyl)phenyl]urea ≥97.0% (by total nitrogen basis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 95.0 area%
  • Purity (with Total Nitrogen):
    min. 97.0 %
  • Melting point:
    160.0 to 164.0 deg-C

Specification Test Results

Related Information

Frequently Bought Together