Diallyl-1,4-cyclohexanedicarboxylate (cis and trans mixture) cis- and trans-mixture ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:20306-22-3
:
Formula:C₁₄H₂₀O₄
MDL Number:MFCD00050452
:252.31 g/mol
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 % (total of isomer)
Specification Test Results
Frequently Bought Together