

N,2-Dimethyl-1,3-propanediamine ≥97.0% (by GC, titration analysis)
Catalog # TCIAD3617-5G
Supplier: TCI
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 97.0 %
- Purity (Nonaqueous Titration):min. 97.0 %
Specification Test Results
About this item
:
:102.18 g/mol
UN:2734
ADR:8,II
:GHS02
:GHS05
CAS Number:1251384-75-4
Formula:C₅H₁₄N₂