

3,4-Dimethylphenylhydrazinium chloride ≥97.0% (by HPLC, titration analysis)
: TCI
:60481-51-8
:MFCD00052270
:C₈H₁₃ClN₂
:172.66 g/mol
:GHS07
:GHS08
- Appearance:White to Amber to Dark purple powder to crystal
- Purity (HPLC):min. 97.0 area%
- Purity (Potassium iodate Method):min. 97.0 %
Frequently Bought Together