
Dichloro(1,5-cyclooctadiene)platinum(II)
Supplier: TCI
Certificates
About this item
CAS Number:12080-32-9
:
Formula:C₈H₁₂Cl₂Pt
MDL Number:MFCD00012413
:374.17 g/mol
EINECS:235-144-5
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Elemental analysis (Carbon):24.9 to 26.4 %
- Chloride:18.0 to 20.0 %
Specification Test Results
Frequently Bought Together