
2',3'-Didehydro-3'-deoxythymidine ≥98.0% (by HPLC, titration analysis)
: TCI
:3056-17-5
:
:C₁₀H₁₂N₂O₄
:MFCD00132921
:224.22 g/mol
:166°C
:GHS08
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
- Specific rotation [a]20/D:-44.0 to -48.0 deg (C=0.69, H2O)
Frequently Bought Together