Order Entry
Netherlands
ContactUsLinkComponent
Dimethyl (1-diazo-2-oxopropyl)phosphonate ≥97.0% (by HPLC)
Dimethyl (1-diazo-2-oxopropyl)phosphonate ≥97.0% (by HPLC)
Supplier:  TCI
Certificates
undefined
Dimethyl (1-diazo-2-oxopropyl)phosphonate ≥97.0% (by HPLC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    Light yellow to Brown clear liquid
  • Purity (HPLC):
    min. 97.0 area%

Specification Test Results

Frequently Bought Together