
6,7-Dihydro-5H-cyclopenta[b]pyridine ≥98.0% (by GC, titration analysis)
: TCI
:533-37-9
:
:C₈H₉N
:MFCD00005933
:119.17 g/mol
:88°C
:67°C
:208-564-1
:GHS07
- Appearance:Colorless to Red to Green clear liquid
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
Frequently Bought Together