Dimethyl-DL-glutamate hydrochloride ≥98.0% (by total nitrogen and titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:13515-99-6
:
Formula:C₇H₁₃NO₄
MDL Number:MFCD00266895
:211.64 g/mol
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (with Total Nitrogen):min. 98.0 %
- Purity (Argentmetric Titration):min. 98.0 %
Specification Test Results
Frequently Bought Together