
3-(Diethoxyphosphoryloxy)-1,2,3-benzotriazin-4(3H)-one ≥98.0% (by HPLC, total nitrogen)
Catalog # TCIAD3262-5G
Supplier: TCI
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
- Melting point:72.0 to 76.0 deg-C
- Solubility in Ethylacetate:almost transparency
Specification Test Results
About this item
:
:299.22 g/mol
:GHS07
CAS Number:165534-43-0
Formula:C₁₁H₁₄N₃O₅P
MDL Number:MFCD01236967