
Disodium-4,4'-bis(2-sulphonatostyryl)biphenyl ≥97.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:27344-41-8
:
Formula:C₂₈H₂₂O₆S₂
MDL Number:MFCD00318874
:562.56 g/mol
EINECS:248-421-0
:GHS07
Specification Test Results
- Appearance:Light yellow to Yellow to Orange powder to crystal
- Purity (HPLC):min. 97.0 area%
Specification Test Results
Frequently Bought Together