Order Entry
Netherlands
ContactUsLinkComponent
1,1-Dichloro-2,2-bis(4-hydroxyphenyl)ethylene ≥98.0% (by GC)
 
1,1-Dichloro-2,2-bis(4-hydroxyphenyl)ethylene ≥98.0% (by GC)
  TCIAD3223-5G
 :  TCI
 :  
 
1,1-Dichloro-2,2-bis(4-hydroxyphenyl)ethylene ≥98.0% (by GC)
  TCIAD3223-5G
 :  TCI
 :  

 

  • Pk:
    5 g
  • Seq:
    3173

 

 

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Melting point:
    213.0 to 219.0 deg-C

 

 

 :
 :281.13 g/mol
 :238-940-0
 :GHS07
 :14868-03-2
 :C₁₄H₁₀Cl₂O₂