2,3-Dimethyl-2,3-butanediamine dihydrochloride ≥98.0% (by total nitrogen and titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:75804-28-3
:
Formula:C₆H₁₆N₂
:189.12 g/mol
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (with Total Nitrogen):min. 98.0 %
- Purity (Argentmetric Titration):min. 98.0 %
Specification Test Results
Frequently Bought Together