Order Entry
Netherlands
ContactUsLinkComponent
 
3,8-Dibromo-1,10-phenanthroline ≥96.0% (by HPLC, titration analysis)
 :  TCI
 
3,8-Dibromo-1,10-phenanthroline ≥96.0% (by HPLC, titration analysis)
 :  TCI

 

  • Appearance:
    White to Light yellow powder to crystal
  • Purity (HPLC):
    min. 96.0 area%
  • Purity (Nonaqueous Titration):
    min. 96.0 %

 

Frequently Bought Together