Order Entry
Netherlands
ContactUsLinkComponent
(S)-(-)-2,2'-Diamino-1,1'-binaphthalene ≥98.0% (by HPLC, titration analysis)
Irritant
(S)-(-)-2,2'-Diamino-1,1'-binaphthalene ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAD3208-200MG
Supplier:  TCI
CAS Number:  
undefined
Irritant
(S)-(-)-2,2'-Diamino-1,1'-binaphthalene ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAD3208-200MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    200 mg
  • Seq:
    2530

Specifications

Specification Test Results

  • Appearance:
    White to Light yellow to Light orange powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Optical purity (LC):
    min. 98.0 ee%
  • Purity (Nonaqueous Titration):
    min. 98.0 %
  • Melting point:
    241.0 to 246.0 deg-C

Specification Test Results

About this item

 :
 :284.362 g/mol
 :GHS07
CAS Number:18531-95-8
Formula:C₂₀H₁₆N₂
MDL Number:MFCD00145204