3,3-Diphenyl-3H-naphtho[2,1-b]pyran ≥98.0% (by HPLC)
TCIAD3197-5G
: TCI
:
- Pk:5 g
- Seq:3173
- Appearance:White to Light yellow powder to crystal
- Purity (HPLC):min. 98.0 area%
- Melting point:161.0 to 165.0 deg-C
:
:334.42 g/mol
:160°C
:4222-20-2
:C₂₅H₁₈O
:MFCD00369541