
Di-tert-butylsilyl-bis(trifluoromethanesulphonate) ≥97.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:85272-31-7
:
Formula:C₁₀H₁₈F₆O₆S₂Si
:440.44 g/mol
Flash Pt:90°C
UN:3265
ADR:8,II
:GHS05
Specification Test Results
- Appearance:Colorless to Light orange to Yellow clear liquid
- Purity (Neutralization titration):min. 97.0 %
Specification Test Results
Frequently Bought Together