
2-Methanesulphonyl-4,6-dimethoxypyrimidine ≥98.0% (by GC)
TCIAD3101-5G
: TCI
:
- Pk:5 g
- Seq:3173
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:128.0 to 132.0 deg-C
:
:218.23 g/mol
:GHS07
:113583-35-0
:C₇H₁₀N₂O₄S
:MFCD00672151