
1,2'-Dinaphthylamine ≥98.0% (by HPLC, total nitrogen), purified by sublimation
Supplier: TCI
Certificates
About this item
CAS Number:4669-06-1
:
Formula:C₂₀H₁₅N
MDL Number:MFCD01632468
:269.35 g/mol
:GHS07
Specification Test Results
- Appearance:White to Yellow to Orange powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
- Melting point:109.0 to 113.0 deg-C
Specification Test Results
Frequently Bought Together