
1,1'-Dinaphthylamine ≥98.0% (by HPLC, total nitrogen)
TCIAD2986-1G
: TCI
:
- Pk:1 g
- Seq:3047
- Appearance:White to Light yellow to Light red powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
- Melting point:110.0 to 115.0 deg-C
:
:257°C
:269.35 g/mol
:GHS07
:737-89-3
:C₂₀H₁₅N