Order Entry
Netherlands
ContactUsLinkComponent
2-(3,4-Dichlorophenyl)ethanol ≥96.0% (by GC)
2-(3,4-Dichlorophenyl)ethanol ≥96.0% (by GC)
Catalog # TCIAD2923-5G
Supplier:  TCI
CAS Number:  
2-(3,4-Dichlorophenyl)ethanol ≥96.0% (by GC)
Catalog # TCIAD2923-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    Colorless to Light yellow to Light orange clear liquid
  • Purity (GC):
    min. 96.0 %

Specification Test Results

About this item

 :
 :191.05 g/mol
Boiling Pt:165°C
CAS Number:35364-79-5
Formula:C₈H₈Cl₂O
MDL Number:MFCD00800673