Order Entry
Netherlands
ContactUsLinkComponent
6,6'-Diamino-2,2'-bipyridyl ≥98.0% (by GC, titration analysis)
Irritant
6,6'-Diamino-2,2'-bipyridyl ≥98.0% (by GC, titration analysis)
Catalog # TCIAD2913-250MG
Supplier:  TCI
CAS Number:  
Irritant
6,6'-Diamino-2,2'-bipyridyl ≥98.0% (by GC, titration analysis)
Catalog # TCIAD2913-250MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    250 mg
  • Seq:
    2590

Specifications

Specification Test Results

  • Appearance:
    White to Yellow to Green powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Purity (Nonaqueous Titration):
    min. 98.0 %
  • Melting point:
    184.0 to 188.0 deg-C

Specification Test Results

About this item

 :
 :186.22 g/mol
 :GHS07
CAS Number:93127-75-4
Formula:C₁₀H₁₀N₄
MDL Number:MFCD04117918