
3,7-Dithia-1,9-nonanedithiol ≥97.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:25676-62-4
:
Formula:C₇H₁₆S₄
MDL Number:MFCD00506918
:228.45 g/mol
:GHS07
Specification Test Results
- Appearance:Light orange to Yellow to Green clear liquid
- Purity (GC):min. 97.0 %
- Purity (Iodometric Titration):min. 97.0 %
Specification Test Results
Frequently Bought Together