(R)-2,2'-Dimethyl-1,1'-binaphthyl ≥98.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:19634-89-0
:
Formula:C₂₂H₁₈
MDL Number:MFCD00048075
:282.39 g/mol
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Melting point:81.0 to 86.0 deg-C
- Specific rotation [a]20/D:-15.0 to -20.0 deg (C=0.5, EtOH (95%))
Specification Test Results
Frequently Bought Together