Order Entry
Netherlands
ContactUsLinkComponent
(S)-Propane-1,2-diamine dihydrochloride ≥98.0% (by titrimetric analysis)
 
(S)-Propane-1,2-diamine dihydrochloride ≥98.0% (by titrimetric analysis)
  TCIAD2828-1G
 :  TCI
 :  
 
(S)-Propane-1,2-diamine dihydrochloride ≥98.0% (by titrimetric analysis)
  TCIAD2828-1G
 :  TCI
 :  

 

  • Pk:
    1 g
  • Seq:
    3047

 

 

  • Appearance:
    White to Almost white powder to crystal
  • Purity (Nonaqueous Titration):
    min. 98.0 %
  • Specific rotation [a]20/D:
    -2.0 to -5.0 deg (C=20, H2O)

 

 

 :
 :147.05 g/mol
 :GHS07
 :19777-66-3
 :C₃H₁₂Cl₂N₂
 :MFCD00050706