trans-1,3-Diphenyl-2-propen-1-ol ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:62668-02-4
:
Formula:C₁₅H₁₄O
MDL Number:MFCD00239426
:210.28 g/mol
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 97.0 %
- Melting point:54.0 to 58.0 deg-C
Specification Test Results
Frequently Bought Together