
Dimethyldioctylammonium bromide ≥97.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:3026-69-5
:
Formula:C₁₈H₄₀BrN
MDL Number:MFCD00082931
:350.43 g/mol
EINECS:221-189-8
:GHS07
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (HPLC) (CAD):min. 95.0 area%
- Purity (Nonaqueous Titration):min. 97.0 %
Specification Test Results
Frequently Bought Together