
1,5-Dimethyl-2-piperidone ≥98.0% (by GC)
Catalog # TCIAD2753-5ML
Supplier: TCI
CAS Number:
Specifications
- Pk:5 mL
- Seq:2210
Specifications
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
About this item
:
:127.19 g/mol
Flash Pt:94°C
:GHS07
CAS Number:86917-58-0
Formula:C₇H₁₃NO
MDL Number:MFCD03701098