threo-5,6-Dodecanediol ≥96.0% (by GC)
Catalog # TCIAD2678-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 96.0 %
- Melting point:46.0 to 50.0 deg-C
- Solubility in Methanol:almost transparency
Specification Test Results
About this item
:
:202.34 g/mol
CAS Number:70859-33-5
Formula:C₁₂H₂₆O₂
MDL Number:MFCD02093463